Orders LinkContactUsLinkComponent
Thread Adapters, Polypropylene
Thread Adapters, Polypropylene
External thread: 64 mm (2.52") (BSI), Internal thread: DIN 61, Yellow/blue
Catalog # 76077-134
Supplier:   BURKLE INC
Supplier #:  0600-1013
Thread Adapters, Polypropylene
Catalog # 76077-134
Supplier:  BURKLE INC
Thread Adapters, Polypropylene
Thread Adapters, Polypropylene
External thread: 64 mm (2.52") (BSI), Internal thread: DIN 61, Yellow/blue
Price
$61.44
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Bürkle offers a broad range of thread adapters to facilitate secure connections between many Bürkle pumps and discharge cocks to a variety of containers
This wide variety of adapters ensures users can find the correct connection.
Specifications
  • Color:
    Yellow/blue
  • Description:
    External thread: 64 mm (2.52") (BSI), Internal thread: DIN 61