Order Entry
United States
Orders LinkContactUsLinkComponent
Thread Adapters, Polypropylene

Thread Adapters, Polypropylene

External thread: 61 mm (2.40"), Internal thread: 3" Coarse, Natural
Catalog # 76077-122
Supplier:   BURKLE INC
Supplier #:  0600-1009
Thread Adapters, Polypropylene
Catalog # 76077-122
Supplier:  BURKLE INC
Thread Adapters, Polypropylene
Thread Adapters, Polypropylene

External thread: 61 mm (2.40"), Internal thread: 3" Coarse, Natural

Price
$69.15
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Bürkle offers a broad range of thread adapters to facilitate secure connections between many Bürkle pumps and discharge cocks to a variety of containers

This wide variety of adapters ensures users can find the correct connection.

Specifications
  • Color:
    Natural
  • Description:
    External thread: 61 mm (2.40"), Internal thread: 3" Coarse