Orders LinkContactUsLinkComponent
EverMark Select Microscope Slides, Azer Scientific
EverMark Select Microscope Slides, Azer Scientific
Frosted slides, color-coded, Orange, 1.0 mm, 75×25 mm (3×1")
Catalog # 89134-541
Supplier #:  EMS200OR
EverMark Select Microscope Slides, Azer Scientific
Catalog # 89134-541
EverMark Select Microscope Slides, Azer Scientific
EverMark Select Microscope Slides, Azer Scientific
Frosted slides, color-coded, Orange, 1.0 mm, 75×25 mm (3×1")
Clearance
$5.98
Sale
Valid until 31/12/9999
Log in for availability
Pack of 72 Case of 1440
Product Details & Documents
Made from high-quality glass, these slides are pre-cleaned with smooth grounded edges.
  • High-quality glass
  • Pre-cleaned
  • Smooth, grounded edges
  • Single-side, color-frosted ends
  • Clipped corners
  • Chemical-resistant
All slides have single-side, color-frosted ends to help avoid sticking and jamming
Adhesive slides feature a permanent positive charge to firmly anchor all types of specimens to the slide. Slides with clipped corners are approved for use with Leica™, Sakura™, Sysmex™, and other automated instruments.
Frosted finish is resistant to common lab chemicals.
Specifications
  • Color:
    Orange
  • Description:
    Frosted slides, color-coded
  • L×W:
    75×25 mm (3×1")
  • Thickness:
    1.0 mm