Orders LinkContactUsLinkComponent
EverMark Select Microscope Slides, Azer Scientific
EverMark Select Microscope Slides, Azer Scientific
Frosted slides, color-coded, Pink, 1.0 mm, 75×25 mm (3×1")
  89033-039
Supplier #:  EMS200P
EverMark Select Microscope Slides, Azer Scientific
  89033-039
EverMark Select Microscope Slides, Azer Scientific
EverMark Select Microscope Slides, Azer Scientific
Frosted slides, color-coded, Pink, 1.0 mm, 75×25 mm (3×1")
$28.78
Pack of 72 Case of 1440
Made from high-quality glass, these slides are pre-cleaned with smooth grounded edges.
  • High-quality glass
  • Pre-cleaned
  • Smooth, grounded edges
  • Single-side, color-frosted ends
  • Clipped corners
  • Chemical-resistant
All slides have single-side, color-frosted ends to help avoid sticking and jamming
Adhesive slides feature a permanent positive charge to firmly anchor all types of specimens to the slide. Slides with clipped corners are approved for use with Leica™, Sakura™, Sysmex™, and other automated instruments.
Frosted finish is resistant to common lab chemicals.
  • Color:
    Pink
  • Description:
    Frosted slides, color-coded
  • L×W:
    75×25 mm (3×1")
  • Thickness:
    1.0 mm