Order Entry
United States
Orders LinkContactUsLinkComponent
VWR® Basic SPP Beard Covers

VWR® Basic SPP Beard Covers

Latex-Free, Blue, XL, 18"
Catalog # 76041-656
Supplier:   VWR International
VWR® Basic SPP Beard Covers
Catalog # 76041-656
Supplier:  VWR International
VWR® Basic SPP Beard Covers
VWR® Basic SPP Beard Covers

Latex-Free, Blue, XL, 18"

Price
$136.14
New Customer Promotional Price
Log in for availability
Case of 1,000
Product Details & Documents

VWR® Basic protection SPP beard covers are manufactured from a spunbonded polypropylene (SPP) fabric that is strong, lightweight, and breathable

  • Economical first-line barrier
  • Non-linting

Elastic band provides secure fit. Available in latex or latex-free formulation. Beard covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.

They protect against dirt, grime, and certain dry particulates in nonhazardous environments. These general-purpose garments are ideal for less critical areas or pre-gowning entry rooms.

Specifications
  • Clothing Size:
    XL
  • Color:
    Blue
  • Description:
    Latex-Free
  • Length:
    18"
  • Tensile Strength:
    >30 (Elongation Percentage)
  • Cat. No.:
    76041-656
  • Particle shedding (Helmke Drum):
    Level II
  • Weight:
    10.0 g/m2
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.