BUFFER PH LOW IONIC STRENGTH 6.97 500ML
Catalog # 103479-528
Supplier: Aqua Solutions
BUFFER PH LOW IONIC STRENGTH 6.97 500ML
Catalog # 103479-528
Supplier: Aqua Solutions
Supplier Number: BE-L1598
Frequently Bought Together
VWR® Weighing Paper
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
Hexane (mixture of isomers) ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Shelf Liners
Supplier: VWR International
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Sleeve Stoppers
Supplier: VWR International
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Supplier: Thermo Fisher Scientific
Reynolds Wrap® Aluminum Foil
Supplier: Control Company
VWR® Dissecting Scissors, Sharp Tip, 4¹/₂"
Supplier: VWR International
neodisher® Detergents, Miele
Supplier: Miele
DuraSeal® Sealing Film, Diversified Biotech
Supplier: Diversified Biotech
PH BUFFER LOW IONIC STRENGTH 4.10
Supplier: Aqua Solutions
Sim Medium
Supplier: VWR International
Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor, Thermo Scientific
Supplier: Thermo Fisher Scientific