EZ RNA METHYLATION KIT THREE-STEP 50PREP
Catalog # 76020-466
Supplier: Zymo Research
EZ RNA METHYLATION KIT THREE-STEP 50PREP
Catalog # 76020-466
Supplier: Zymo Research
Supplier Number: R5001
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Corning® Treated Culture Dishes, Corning
Supplier: Corning
High Sensitivity DNA ScreenTape Analysis
Supplier: Agilent Technologies