YEAST PLASMID MINIPREP KIT ZYMOPREP-96 8
Catalog # 76020-318
Supplier: Zymo Research
YEAST PLASMID MINIPREP KIT ZYMOPREP-96 8
Catalog # 76020-318
Supplier: Zymo Research
Supplier Number: D2007
Frequently Bought Together
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
ART® and Hydrologix (HLT) SoftFit-L™ Pipette Tips for Rainin® LTS Pipettors, Molecular BioProducts
Supplier: Thermo Scientific
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Dithiothreitol (DTT, Cleland's reagent) ≥99.0%, Molecular Grade
Supplier: Promega Corporation
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics