YEAST PLASMID MINIPREP KIT ZYMOPREP-96 4
Catalog # 76020-316
Supplier: Zymo Research
YEAST PLASMID MINIPREP KIT ZYMOPREP-96 4
Catalog # 76020-316
Supplier: Zymo Research
Supplier Number: D2006
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
VWR® Weighing Paper
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
CONJUGATE STREPTAVIDIN HRP E-COLI 1ML
Supplier: Southern Biotechnology