Polystyrene Reagent Reservoirs
Reagent Reservoir - Divided, 50 ml, 28 mm, 146 mm, 83 mm, Sterile
470228-474
: P8051-5S_WARDS
Standard 4-channel, 8-channel, 12-channel and various special purpose pipettes are all compatible with these disposable, polystyrene reservoirs. To facilitate easy, dripless pouring, all versions have independent spouts in each corner. Bottoms are formed with a sharply tapered 'V' shape to minimize waste and reagent consumption.
- Graduated
- Anti-static
- Sharp "V" Bottoms
- Pouring spout on all 4 corners
- Divided options are available
- All reservoirs are RNAse- and DNAse-free
- Sterility:Sterile
- Width:83 mm
- Capacity:50 ml
- Description:Reagent Reservoir - Divided
- Height:28 mm
- Length:146 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



