Order Entry
United States
Orders LinkContactUsLinkComponent
1,1,2,2-Tetrachloroethane

1,1,2,2-Tetrachloroethane

Steel can, 20 L
Catalog # 76014-922
Supplier #:  T1018-20LT
1,1,2,2-Tetrachloroethane
Catalog # 76014-922
CAS Number:  79-34-5
1,1,2,2-Tetrachloroethane
1,1,2,2-Tetrachloroethane

Steel can, 20 L

CAS Number:  79-34-5
Price
$3,095.62
Log in for availability
Each (20l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

1,1,2,2-Tetrachloroethane, also known as Acetylene tetrachloride, is a chlorinated derivative of ethane with a very high solvent power.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

  • CAS Number:79-34-5
  • Formula:Cl₂HCCHCl₂
  • MDL Number:MFCD00000848
  • Molecular Weight:167.85 g/mol
  • Density:1.586
  • Storage Temperature:Ambient
  • UN:1702
  • ADR:6.1,II

Documents

Specifications

Specifications

  • Pack type:
    Steel can
  • Size:
    20 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.