Order Entry
United States
Orders LinkContactUsLinkComponent
Human Recombinant Matriptase 2

Human Recombinant Matriptase 2

500 U
Catalog # 76014-124
Supplier:   Enzo Life Sciences
Supplier #:  201-752-0500
Human Recombinant Matriptase 2
Catalog # 76014-124
Supplier:  Enzo Life Sciences
CAS Number:  
Human Recombinant Matriptase 2
Human Recombinant Matriptase 2

500 U

CAS Number:  
Price
$661.63
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Produced in Drosophila Schneider 2 (S2) cells. Recombinant human matriptase-2 (aa 78-811) is fused at the C-terminus to a V5-His-tag.

  • Trypsin-like serine protease.

Matriptase-2 is a potent negative regulator of hepcidin.

Specifications

Specifications

  • Size:
    500 U
  • Cat. No.:
    76014-124
  • Enzyme type:
    Recombinant
  • Form:
    Liquid
  • Formulation:
    In 50 mM HEPES, pH 9.0, containing 400 mM sodium chloride and 20% glycerol.
  • Enzyme Activity:
    ~2.5 U/µl. One unit is defined at the amount of enzyme that releases 1 nmol of AMC per min. at 37 °C in the presence of 50 µM Boc-Gln-Ala-Arg-AMC.
  • Concentration:
    100 nM
  • Species:
    Human
  • Enzyme Name:
    Matriptase 2

Specifications