Spectroscopy Light Sources
Power Supply for Light Source, 220V
Catalog # 470228-030
Jump to:
Discontinued
Product Details & Documents
Various type of light sources for spectroscopy experiments.
- Predicatable spectral lines
- Includes a closable shield block
Perfect for spectroscopy and other optics experiments. Sodium and mercury light sources have predictable and unique spectral lines which make them perfect for calibrating your spectrometers or other equipment. Each lamp house includes a closable shield to block out light when it is not needed.
Ordering information: The power supply, available separately, can power one lamp at a time.
Specifications
- Description:Power Supply for Light Source, 220V