COLUMN ZYMO-SPIN IIICG DNA/RNA BINDING
Catalog # 76004-148
Supplier: Zymo Research
COLUMN ZYMO-SPIN IIICG DNA/RNA BINDING
Catalog # 76004-148
Supplier: Zymo Research
Supplier Number: C1006-250-G
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® PCR Strip Tubes & Caps 8-Strip
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Corning® Trypsin
Supplier: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Histoware™ Prefilled Specimen Containers, Caplugs
Supplier: Caplugs
ZR BashingBead Lysis/Filtration Tubes, Zymo Research
Supplier: Zymo Research
Electroporation Cuvettes, Bulldog Bio
Supplier: Bulldog Bio
Ethanol ≥70%, KOPTEC USP, Biotechnology Grade (140 Proof)
Supplier: Decon Labs