Orders LinkContactUsLinkComponent
Dual Channel Function/Arbitrary Waveform Generator
Dual Channel Function/Arbitrary Waveform Generator
60 MHz Dual Channel Function/Arbitrary Waveform Generator
Catalog # 470227-650
Supplier:   B & K Precision
Supplier #:  4055B
Dual Channel Function/Arbitrary Waveform Generator
Catalog # 470227-650
Supplier:  B & K Precision
Dual Channel Function/Arbitrary Waveform Generator
Dual Channel Function/Arbitrary Waveform Generator
60 MHz Dual Channel Function/Arbitrary Waveform Generator
Price
$1,573.99
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
More power for higher frequency waveform generation.
  • 14-bit, 150 MSa/s, 16k point arbitrary waveform generator
  • Two independent channels with individual output On/Off buttons
  • Convenient channel copy, track and combine functions
  • Synchronize the phase of both channels with the push of a button
  • Low-jitter square wave generation for simulating reliable clock signals, generating triggers, or validating serial data buses
  • Large 4.3-inch LCD color display
The Dual Channel Function/Arbitrary Waveform Generators are capable of generating stable and precise sine, square, triangle, pulse, and arbitrary waveforms. With an easy-to-read color display and intuitive user interface with numeric keypad, these instruments offer plenty of features including linear/logarithmic sweep, built-in counter, extensive modulation and triggering capabilities, a continuously variable DC offset, and a high performance 14-bit, 150 MSa/s arbitrary waveform generator. CH1 and CH2 outputs can both be varied from 0 to 10 Vpp into 50 ohms (up to 20 Vpp into open circuit).
Ordering information: See the manual for more detailed specifications.
Specifications
  • Description:
    60 MHz Dual Channel Function/Arbitrary Waveform Generator