ASSAY HUMAN IFN-Y ALP STRIPS 2 PLATES
Catalog # MSPP-34204AST2
Supplier: MABTECH INC MS
ASSAY HUMAN IFN-Y ALP STRIPS 2 PLATES
Catalog # MSPP-34204AST2
Supplier: MABTECH INC MS
Supplier Number: 3420-4AST-2
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
VWR® Weighing Paper
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
Hexane (mixture of isomers) ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Sleeve Stoppers
Supplier: VWR International
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Supplier: Thermo Fisher Scientific
Reynolds Wrap® Aluminum Foil
Supplier: Antyila Scientific
Polystyrene and Polypropylene Test Tubes, 12×75 mm, Globe Scientific
Supplier: Globe Scientific
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics