Orders LinkContactUsLinkComponent
Micro Scissors, Sklar
Micro Scissors, Sklar
Micro Scissors, Round Handle, 178 mm (7"), Stainless steel, Sharp/Sharp, Smooth, Straight
Catalog # 75996-324
Supplier:   Sklar
Supplier #:  98-7837
Micro Scissors, Sklar
Catalog # 75996-324
Supplier:  Sklar
Micro Scissors, Sklar
Micro Scissors, Sklar
Micro Scissors, Round Handle, 178 mm (7"), Stainless steel, Sharp/Sharp, Smooth, Straight
Price
$1,876.02
New Customer Promotional Price
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sklar's® micro scissors are used for the dissection of tissues, blood vessels and nerves.
  • Reusable
  • Nonsterile
Available with a flat or round handle. The straight scissors may be used to cut sutures and trim nerve endings. Curved will dissect vessels and nerves. In addition to the tips, the sides may be used for dissection with the tips closed, similar to using forceps.
Specifications
  • Description:
    Micro Scissors, Round Handle
  • Length:
    178 mm (7")
  • Material:
    Stainless steel
  • Curvature:
    Straight
  • Blade type:
    Sharp/Sharp, Smooth