Order Entry
United States
Orders LinkContactUsLinkComponent
Buffer Solution, Color-Coded, Fluka™, Honeywell Research Chemicals

Buffer Solution, Color-Coded, Fluka™, Honeywell Research Chemicals

±0.05, Borate, 20 °C, Plastic bottle, 6× 500 ml, 10.0, Violet
Catalog # BJ33668-500ML
Buffer Solution, Color-Coded, Fluka™, Honeywell Research Chemicals
Catalog # BJ33668-500ML
Supplier:  
CAS Number:  
Buffer Solution, Color-Coded, Fluka™, Honeywell Research Chemicals

±0.05, Borate, 20 °C, Plastic bottle, 6× 500 ml, 10.0, Violet

CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These buffer solutions are color-coded for easy identification and the pH 4 and 7 version have fungicide additive.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Color:
    Violet
  • Size:
    6× 500 ml
  • pH buffer type:
    Borate
  • Reference temperature:
    20 °C
  • pH:
    10.0
  • pH accuracy:
    ±0.05

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.