Order Entry
United States
Orders LinkContactUsLinkComponent
 

Buffer Solution, Color-Coded, Fluka™, Honeywell Research Chemicals

±0.02, Citrate, 20 °C, Plastic bottle, 6× 500 ml, 4.0, Red
  BJ33665-500ML
Buffer Solution, Color-Coded, Fluka™, Honeywell Research Chemicals
  BJ33665-500ML
 :  
 :  
Buffer Solution, Color-Coded, Fluka™, Honeywell Research Chemicals

±0.02, Citrate, 20 °C, Plastic bottle, 6× 500 ml, 4.0, Red

 :  
 

These buffer solutions are color-coded for easy identification and the pH 4 and 7 version have fungicide additive.

 

 

  • Pack type:
    Plastic bottle
  • Color:
    Red
  • Size:
    6× 500 ml
  • pH buffer type:
    Citrate
  • Reference temperature:
    20 °C
  • pH:
    4.0
  • pH accuracy:
    ±0.02

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.