PCR Plates, Watson Bio Lab
96, Semi-Skirted, 0,1 ml
Catalog # 75982-970
Supplier: Watson Bio Lab
Supplier #: 137-675C
Jump to:
Product Details & Documents
The wells are uniformly thin, which enables PCR with superior heat exchange.
- Precast Demarcation Lines
- Thin Wall
- High Transparency
- Nuclease-free Certified
- Autoclavable Polypropylene Construction
These PCR plates feature autoclavable polypropylene construction and precast demarcation lines. Users can cut the plates as needed.
Specifications
- No. of wells:96
- Skirt:Semi-Skirted
- Well volume:0,1 ml