RECOMBINANT HUMAN INTERLEUKIN-4 10UG
Catalog # MSPP-RCYECHIL4
Supplier: Invivogen
RECOMBINANT HUMAN INTERLEUKIN-4 10UG
Catalog # MSPP-RCYECHIL4
Supplier: Invivogen
Supplier Number: RCYEC-HIL4
Frequently Bought Together
Vi-CELL BLU Reagent Kits, Beckman Coulter
Supplier: Beckman Coulter
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
Nunc-Immuno™ MicroWell™ 96-Well Plates, Thermo Scientific
Supplier: Thermo Scientific
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
Disposable PD-10 Desalting Columns, Cytiva
Supplier: Cytiva
FlashGel™ DNA, RNA and Recovery Systems
Supplier: Lonza
BUFFER TBS BLOCKING READY-TO-USE 500ML
Supplier: Li-Cor
Human TruStain FcX™, Fc Receptor Blocking Solution, BioLegend
Supplier: Biolegend
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
Laemmli SDS sample buffer, reducing (6X), Thermo Scientific
Supplier: Thermo Scientific Chemicals