Order Entry
United States
Orders LinkContactUsLinkComponent
Mouse Recombinant CCL8

Mouse Recombinant CCL8

20 µg
Catalog # 75933-896
Supplier #:  010-001-V75-0020
Mouse Recombinant CCL8
Catalog # 75933-896
CAS Number:  
Mouse Recombinant CCL8

20 µg

CAS Number:  
Price
$356.66
Log in for availability
Each (20µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recombinant Mouse CCL8 control protein

  • Control Protein
  • PCA Assays
  • ImmunoBlot

Monocyte Chemotactic protein 2 (MCP-2), also known as CCL8, is implicated in allergic responses through its ability to activate mast cells, eosinophils, and basophils. MCP-2 is known to signal through several G protein-coupled receptors including, CCR1, CCR2B, and CCR5. Recombinant mouse MCP-2 is a non-glycosylated protein, containing 74 amino acids, with a molecular weight of 8.5 kDa.

Monocyte Chemotactic Protein-2 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Monocyte Chemotactic Protein-2 in immunological assays.

Specifications

Specifications

  • Size:
    20 µg
  • Cat. No.:
    75933-896
  • Shipping Temperature:
    Ambient
  • Protein/Peptide Type:
    Recombinant
  • Protein/Peptide Name:
    CCL8
  • Species:
    Mouse

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.