Orders LinkContactUsLinkComponent
Test Kits, COD, TNT plus®
Test Kits, COD, TNT plus®
COD TNTplus™, HR (EPA, digestion required), 25 Tubes/pack, Reactor digestion, 20 - 1500 mg/L COD
Catalog # 75919-300
Test Kits, COD, TNT plus®
Catalog # 75919-300
Supplier:  
CAS Number:  
Test Kits, COD, TNT plus®
Test Kits, COD, TNT plus®
COD TNTplus™, HR (EPA, digestion required), 25 Tubes/pack, Reactor digestion, 20 - 1500 mg/L COD
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
VIAL COD HR+ 0-15 000MG/L 25 T Catalog #97009-964
Product 97009-964 is a replacement for 75919-300 and has the same packaging and specifications, but may be a different quantity.
REAGENT SET NITRAVER X 50TEST Catalog #104088-658
Product 104088-658 is a replacement for 75919-300 and has the same packaging and specifications, but may be a different quantity.
REAGENT IRON SOLUTIN F TRACE LEVEL 500ML Catalog #104088-200
Product 104088-200 is a replacement for 75919-300 and has the same packaging and specifications, but may be a different quantity.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are color coded by parameter and range
  • No reagent blank necessary
TNTplus™ works only with DR 6000™ UV-VIS, DR 3900™ Benchtop, and DR 1900™ Portable spectrophotometers.
Specifications
Specifications
  • Chemical method:
    Reactor digestion
  • Description:
    COD TNTplus™, HR (EPA, digestion required)
  • Size:
    25 Tubes/pack
  • Range:
    20 - 1500 mg/L COD