Orders LinkContactUsLinkComponent
Test Kits, COD, TNT plus®
COD TNTplus™, LR (EPA, digestion required), 150 Tubes/pack, Reactor digestion, 3 - 150 mg/L COD
  75919-248
Test Kits, COD, TNT plus®
  75919-248
 :  
 :  
Test Kits, COD, TNT plus®
Test Kits, COD, TNT plus®
COD TNTplus™, LR (EPA, digestion required), 150 Tubes/pack, Reactor digestion, 3 - 150 mg/L COD
 :  
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are color coded by parameter and range
  • No reagent blank necessary
TNTplus™ works only with DR 6000™ UV-VIS, DR 3900™ Benchtop, and DR 1900™ Portable spectrophotometers.
  • Chemical method:
    Reactor digestion
  • Description:
    COD TNTplus™, LR (EPA, digestion required)
  • Size:
    150 Tubes/pack
  • Range:
    3 - 150 mg/L COD