Test Kits, COD, TNT plus®
Product 104087-808 is similar to 75919-246 but manufacturer and specifications may vary.
Product 89212-756 is similar to 75919-246 but manufacturer and specifications may vary.
Product Details & Documents
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process
- Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
- Barcode helps the instrument to automatically detect and run the correct method
- Barcode allows spectrophotometer to detect reagent lot number and expiry date
- Vials have flat bottom to improve handling
- Vials are color coded by parameter and range
- No reagent blank necessary
TNTplus™ works only with DR 6000™ UV-VIS, DR 3900™ Benchtop, and DR 1900™ Portable spectrophotometers.
Documents
Specifications
Specifications
- Chemical method:Reactor digestion
- Description:COD TNTplus™, LR (EPA, digestion required)
- Size:25 Tubes/pack
- Range:3 - 150 mg/L COD