Orders LinkContactUsLinkComponent
Test Kits, COD, TNT plus®
Test Kits, COD, TNT plus®
COD TNTplus™, ULR (digestion required), 25 Tubes/pack, Reactor digestion, 1 - 60 mg/L COD
Catalog # 75919-242
Test Kits, COD, TNT plus®
Catalog # 75919-242
Supplier:  
CAS Number:  
Test Kits, COD, TNT plus®
Test Kits, COD, TNT plus®
COD TNTplus™, ULR (digestion required), 25 Tubes/pack, Reactor digestion, 1 - 60 mg/L COD
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
COD TNTPLUS 1.0 60 MG L 25TESTS F DR3900 Catalog #104088-654
Product 104088-654 is a replacement for 75919-242 but varies by Brand, Packaging or Quantity.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are color coded by parameter and range
  • No reagent blank necessary
TNTplus™ works only with DR 6000™ UV-VIS, DR 3900™ Benchtop, and DR 1900™ Portable spectrophotometers.
Specifications
Specifications
  • Chemical method:
    Reactor digestion
  • Description:
    COD TNTplus™, ULR (digestion required)
  • Size:
    25 Tubes/pack
  • Range:
    1 - 60 mg/L COD