Orders LinkContactUsLinkComponent
Prosolv® Recycling System and Components, Justrite®
Prosolv® Recycling System and Components, Justrite®
Prosolv® Gas Cylinder Recycling System
Catalog # 75870-996
Supplier:   Justrite
Supplier #:  28190
Prosolv® Recycling System and Components, Justrite®
Catalog # 75870-996
Supplier:  Justrite
Prosolv® Recycling System and Components, Justrite®
Prosolv® Gas Cylinder Recycling System
Price
$1,541.39
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Safely and economically convert gas cylinders into recyclable steel containers.
  • Prosolv® System removes the sealing sleeve and internal valve stem from propane, MAPP, and 0.75" male threaded calibration gas cylinders
  • Empties and filters propellants from cylinders, converting them into non-hazardous recyclable material
  • Not for use with oxygen bottles
Spent propane canisters are considered hazardous waste while still under pressure, but once they are safely vented and relieved of pressure they can be recycled as scrap steel. The Prosolv® System safely recycles depressurized propane cylinders, eliminating an entire category of hazardous waste from the waste stream.
Certifications: EPA Compliant.
Accessories information: System includes vent device, carbon filter, antistatic wire, safety goggles, sleeve removal tool, and 40 certification tags.
Specifications
  • Description:
    Prosolv® Gas Cylinder Recycling System