Order Entry
United States
Orders LinkContactUsLinkComponent
Trypsin Kit, Electron Microscopy Science

Trypsin Kit, Electron Microscopy Science

Trypsin Kit Large
Catalog # 103305-614
Supplier #:  64142-02
Trypsin Kit, Electron Microscopy Science
Catalog # 103305-614
CAS Number:  
Trypsin Kit, Electron Microscopy Science

Trypsin Kit Large

CAS Number:  
Price
$236.98
Log in for availability
Kit of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Trypsin is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies.

  • Use as antigen retriever for some antibodies in IHC
  • Please refer to primary antibody protocol for IHC
  • Supplied as concentrated form along with buffer

In IHC most commonly used fixative like formalin; mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC. Trypsin digestion improves immunoreactivity of some antigens in FFPE tissue sections. For cytokeratin clone AE3 and AE1/AE3, this enzyme works much better than boiling the tissue with citrate buffer.

Delivery information: Small kit contains (15 mL) Reagent B Buffer and (2 mL) Reagent E (Enzyme).
Large kit contains (100 mL) Reagent B Buffer and (10 mL) Reagent E (Enzyme).

Specifications

Specifications

  • Description:
    Trypsin Kit Large
  • Cat. No.:
    103305-614
  • Storage Temperature:
    2-8°C. DO NOT FREEZE.

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.