Orders LinkContactUsLinkComponent
Trypsin Kit, Electron Microscopy Science
Trypsin Kit, Electron Microscopy Science
Trypsin Kit Small
Catalog # 103303-052
Supplier #:  64142-01
Trypsin Kit, Electron Microscopy Science
Catalog # 103303-052
CAS Number:  
Trypsin Kit, Electron Microscopy Science
Trypsin Kit Small
CAS Number:  
Price
$123.21
Log in for availability
Pack of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Trypsin is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies.
  • Use as antigen retriever for some antibodies in IHC
  • Please refer to primary antibody protocol for IHC
  • Supplied as concentrated form along with buffer
In IHC most commonly used fixative like formalin; mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC. Trypsin digestion improves immunoreactivity of some antigens in FFPE tissue sections. For cytokeratin clone AE3 and AE1/AE3, this enzyme works much better than boiling the tissue with citrate buffer.
Delivery information: Small kit contains (15 mL) Reagent B Buffer and (2 mL) Reagent E (Enzyme).
Large kit contains (100 mL) Reagent B Buffer and (10 mL) Reagent E (Enzyme).
Specifications
Specifications
  • Description:
    Trypsin Kit Small