Orders LinkContactUsLinkComponent
Cell Stretcher, Electron Microscopy Sciences
Membrane for Cell Stretcher
  103302-220
Cell Stretcher, Electron Microscopy Sciences
  103302-220
Cell Stretcher, Electron Microscopy Sciences
Cell Stretcher, Electron Microscopy Sciences
Membrane for Cell Stretcher
$100.07
Each
Cell Stretcher CS-10 Series is a device for simultaneous live cell imaging with motion compensation for uni-axial mechanical straining or compression.
  • Cells can be imaged even during stretch
  • Stretching chamber allows use of commercially available PDMS sheets
  • Cyclic protocols with different characteristics (linear, sinusoidal, sigmoidal) can be programmed
  • Position of the membrane chamber along the stretch axis can be changed
This compact cell strain device makes it possible to perform life cell imaging studies where a rapid image acquisition is required, such as observing changes of the intracellular Ca2+ concentration when cells are mechanically stimulated.
  • Description:
    Membrane for Cell Stretcher