7-AAD SOLN 488-650NM 200TST FOR FC ASSAY
Catalog # 75843-404
Supplier: BIOGEMS INTERNATIONAL INC.
7-AAD SOLN 488-650NM 200TST FOR FC ASSAY
Catalog # 75843-404
Supplier: BIOGEMS INTERNATIONAL INC.
Supplier Number: 61410-00-200
Frequently Bought Together
Corning® Internal Thread Cryogenic Vials, Corning
Supplier: Corning
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
Anti-Calbindin D-28K Chicken Polyclonal Antibody
Supplier: Novus Biologicals