Assorted Plastic Pulleys
Assorted Pulleys, Pack of 100
Catalog # 470224-022
Supplier: SI Manufacturing
Supplier #: 50030
Jump to:
Product Details & Documents
This package of plastic pulleys contains 25 each of 20, 30 and 40 mm diameter pulleys with 3.2 mm bores and one 10 mm pulley with a push fit to a 2 mm motor shaft.
: Pack of 100.
: CHOKING HAZARD- small parts not for children under 8 years.
Specifications
- Description:Assorted Pulleys, Pack of 100