Orders LinkContactUsLinkComponent
BOD Bottles
BOD Bottles
300 ml
Catalog # 470223-794
Supplier #:  BOD300
BOD Bottles
Catalog # 470223-794
BOD Bottles
BOD Bottles
300 ml
Price
$20.70
Log in for availability
Each Pack of 24
Product Details & Documents
For biochemical oxygen demand testing.
  • Glass bottles
  • Bottles are unnumbered
  • Feature penny head stopper
BOD Bottles are designed for incubating diluted samples of clean water, waste water, effluent water, and polluted water during the five-day biochemical oxygen demand (BOD) test.
Specifications
  • Capacity:
    300 ml