Order Entry
United States
Orders LinkContactUsLinkComponent
RapidClean, Advansta

RapidClean, Advansta

RapidClean Resin, 1 mL
Catalog # 103254-760
Marketsource
Supplier:   ADVANSTA INC MS
Supplier #:  R-14011-B10
RapidClean, Advansta
Catalog # 103254-760
Supplier:  ADVANSTA INC MS
Marketsource
CAS Number:  
RapidClean, Advansta

RapidClean Resin, 1 mL

CAS Number:  
Price
$289.62
Log in for availability
Each (1ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Rapid clean is the fast and easy way to remove protein from RNA and DNA samples.

  • Complete removal of proteins from aqueous solutions of nucleic acids
  • Protocol takes less than 5 min
  • Provides non-toxic alternative to phenol-chloroform extraction
  • Nucleic acid recovery rates >95%
  • Recovers nucleic acids from 5 to greater than 40 kb
  • Suitable for DNA, RNA, cDNA, microRNA, and oligonucleotides

In less than 5 minutes, completely remove protein from aqueous solutions of single- and double-stranded nucleic acids.

RapidClean is a novel affinity resin designed to remove all protein from aqueous solutions of nucleic acids, providing an alternative to hazardous and lengthy phenol-chloroform procedures. The RapidClean resin binds and removes protein in a 5-minute protocol that combines convenience, speed, and nucleic acid recovery rates in excess of 95%.

 : Kit includes RapidClean resin and spin filters.

Specifications

Specifications

  • Description:
    RapidClean Resin
  • Size:
    1 mL

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.