Orders LinkContactUsLinkComponent
RapidClean, Advansta
RapidClean, Advansta
RapidClean Protein Removal Kit, 25 rxns
Catalog # 103254-756
Marketsource
Supplier:   ADVANSTA INC MS
Supplier #:  K-01001-025
RapidClean, Advansta
Catalog # 103254-756
Supplier:  ADVANSTA INC MS
Marketsource
CAS Number:  
RapidClean, Advansta
RapidClean Protein Removal Kit, 25 rxns
CAS Number:  
Price
$446.54
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rapid clean is the fast and easy way to remove protein from RNA and DNA samples.
  • Complete removal of proteins from aqueous solutions of nucleic acids
  • Protocol takes less than 5 min
  • Provides non-toxic alternative to phenol-chloroform extraction
  • Nucleic acid recovery rates >95%
  • Recovers nucleic acids from 5 to greater than 40 kb
  • Suitable for DNA, RNA, cDNA, microRNA, and oligonucleotides
In less than 5 minutes, completely remove protein from aqueous solutions of single- and double-stranded nucleic acids.
RapidClean is a novel affinity resin designed to remove all protein from aqueous solutions of nucleic acids, providing an alternative to hazardous and lengthy phenol-chloroform procedures. The RapidClean resin binds and removes protein in a 5-minute protocol that combines convenience, speed, and nucleic acid recovery rates in excess of 95%.
Ordering information: Kit includes RapidClean resin and spin filters.
Specifications
Specifications
  • Description:
    RapidClean Protein Removal Kit
  • Size:
    25 rxns