Orders LinkContactUsLinkComponent
RapidClean, Advansta
RapidClean Protein Removal Kit, 10 rxns
  103254-754
Marketsource
 :  K-01001-010
RapidClean, Advansta
  103254-754
Marketsource
 :  
RapidClean, Advansta
RapidClean Protein Removal Kit, 10 rxns
 :  
$221.54
Each
Rapid clean is the fast and easy way to remove protein from RNA and DNA samples.
  • Complete removal of proteins from aqueous solutions of nucleic acids
  • Protocol takes less than 5 min
  • Provides non-toxic alternative to phenol-chloroform extraction
  • Nucleic acid recovery rates >95%
  • Recovers nucleic acids from 5 to greater than 40 kb
  • Suitable for DNA, RNA, cDNA, microRNA, and oligonucleotides
In less than 5 minutes, completely remove protein from aqueous solutions of single- and double-stranded nucleic acids.
RapidClean is a novel affinity resin designed to remove all protein from aqueous solutions of nucleic acids, providing an alternative to hazardous and lengthy phenol-chloroform procedures. The RapidClean resin binds and removes protein in a 5-minute protocol that combines convenience, speed, and nucleic acid recovery rates in excess of 95%.
 : Kit includes RapidClean resin and spin filters.
  • Description:
    RapidClean Protein Removal Kit
  • Size:
    10 rxns