MICROPLATE QUICKPLEX 96W HIGH BIND PK10
MSPP-L55XB3
Frequently Bought Together
Reaction Tubes with Soft Push Cap
: SARSTEDT INC
Microcentrifuge Screw Caps Closures for Micrewtubes™ Centrifuge Tubes, Simport
: Simport Scientific
VWR® Screw Cap Microtubes with Customizable Caps
: VWR International
Micrewtube™ Microcentrifuge Tubes, Simport Scientific
: Simport Scientific
GeneMate Screw Cap Microtubes and Caps, Scientific Specialties
: VWR International
VWR® Disposable Microcentrifuge Tubes
: VWR International
MµlTI™ SafeSeal™ Microcentrifuge Tubes, Sorenson BioScience
: Sorenson Bioscience
RECOMBINANT HUMAN INTERLEUKIN-4 10UG
: Invivogen
Certified Free and Sterile Ultracentrifugation Tubes, Beckman Coulter
: Beckman Coulter
Tubes with False Bottoms
: SARSTEDT INC
Certified Microcentrifuge Tubes in Self-Standing Bags, Globe Scientific
: Globe Scientific