Order Entry
United States
Orders LinkContactUsLinkComponent
D-(-)-Mannitol, BiotechGrade

D-(-)-Mannitol, BiotechGrade

Poly pail, 2.5 kg
Catalog # 75811-566
Supplier #:  M1128-2.5KG
D-(-)-Mannitol, BiotechGrade
Catalog # 75811-566
CAS Number:  69-65-8
D-(-)-Mannitol, BiotechGrade
D-(-)-Mannitol, BiotechGrade

Poly pail, 2.5 kg

CAS Number:  69-65-8
Price
$1,208.06
Log in for availability
Each (2,500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    Fine,White,Cryst
  • Assay (C₆H₁₄O₆; Dried Basis):
    96.0 - 101.5 %
  • Appearance of Solution (2 G in 20 ML):
    Clear and Colorl
  • Melting Range:
    164 - 169°C
  • Specific Rotation, [A]D25:
    +137° to +145°

Specifications

  • Pack type:
    Poly pail
  • Size:
    2.5 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.