D-(-)-Mannitol, BiotechGrade
Poly pail, 12 kg
Catalog # 75811-564
Supplier: Spectrum Chemical Mfg. Corp.
Supplier #: M1128-12KG
D-(-)-Mannitol, BiotechGrade
Catalog # 75811-564
Supplier: Spectrum Chemical Mfg. Corp.
CAS Number: 69-65-8
Jump to:
Product Details & Documents
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Storage Temperature:Ambient
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Appearance:Fine,White,Cryst
- Assay (C₆H₁₄O₆; Dried Basis):96.0 - 101.5 %
- Appearance of Solution (2 G in 20 ML):Clear and Colorl
- Melting Range:164 - 169°C
- Specific Rotation, [A]D25:+137° to +145°
Specifications
- Pack type:Poly pail
- Size:12 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



