ACETONITRILE HPLC GRADE 75-05-8 LIQ 4L
Catalog # 103193-964
Supplier: Aqua Solutions
ACETONITRILE HPLC GRADE 75-05-8 LIQ 4L
Catalog # 103193-964
Supplier: Aqua Solutions
Supplier Number: A1520-4LSS
Frequently Bought Together
VWR® Disposable Petri Dishes
Supplier: VWR International
Toluene ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
Falcon® Cell Culture Flasks, Corning
Supplier: Corning
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate 98%
Supplier: Ambeed