NITRIC ACID W/W (3.930+/-0.005) NIST 4L
Catalog # 103193-820
Supplier: Aqua Solutions
NITRIC ACID W/W (3.930+/-0.005) NIST 4L
Catalog # 103193-820
Supplier: Aqua Solutions
Supplier Number: ALC027A
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Methanol, anhydrous ≥99.8% (by GC, corrected for water content), BAKER ANANYZED™ ACS for Karl Fischer titration, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC