ROTOR ANGLE 24-PLACE FOR MIKRO 200 CENT
Catalog # 75801-708
Supplier: HETTICH INSTRUMENTS
ROTOR ANGLE 24-PLACE FOR MIKRO 200 CENT
Catalog # 75801-708
Supplier: HETTICH INSTRUMENTS
Supplier Number: 2428
Frequently Bought Together
AiroSensor Temperature Sensor, −40 to +70 °C
Supplier: SenseAnywhere
H-B® Easy Read® Environmentally Friendly, General Purpose Liquid-In-Glass Thermometers
Supplier: Bel-Art Products
Red Liquid Filled Wall Thermometer
Supplier: Taylor Precision Products
Frontier™ 5000 Multi-Pro Centrifuges, 230 V
Supplier: Ohaus
ASTM-Like Non-Mercury Thermometers, Thermco
Supplier: Thermco
Accessories for Excellence Level, XPR/XSR Series Analytical Balances
Supplier: Mettler Toledo
JS-4.3 Swinging Bucket Rotor, Beckman Coulter®
Supplier: Beckman Coulter
Modular Disk Adapters for SX4750, SX4750A, and JS-4.3 Rotors, Beckman Coulter®
Supplier: Beckman Coulter
Rotors for Vacufuge Plus
Supplier: Eppendorf
Accessories for SX4750 and SX4750A Swinging Bucket Rotors, Beckman Coulter
Supplier: Beckman Coulter
Rsa I, Restriction Enzymes, Promega
Supplier: Promega Corporation
Eppendorf® 5430 R Refrigerated Microcentrifuges
Supplier: Eppendorf
TEMP HUMIDITY DATA LOGGER
Supplier: Thermco
Rotors for Centrifuges, 5810/5810 R
Supplier: Eppendorf
Pico™ and Fresco™ Microcentrifuges, Thermo Scientific
Supplier: Thermo Fisher Scientific
Eppendorf® 5430 Microcentrifuges
Supplier: Eppendorf
Accessories for VWR® Data Logging Ethernet Thermometers Compatible with TraceableLIVE® Cloud Service
Supplier: VWR International
Large-Digit Triple Display Thermometers, Thermco
Supplier: Thermco