Orders LinkContactUsLinkComponent
Mesh Stack and Hang Bins, Quantum Storage Systems
Mesh Stack and Hang Bins, Quantum Storage Systems
Exterior: 14 1/2 × 16 1/4 × 7"
Interior: 14 × 15 3/4 × 6 3/4"
Catalog # 10804-545
Supplier #:  QMB550C
Mesh Stack and Hang Bins, Quantum Storage Systems
Catalog # 10804-545
Mesh Stack and Hang Bins, Quantum Storage Systems
Mesh Stack and Hang Bins, Quantum Storage Systems
Exterior: 14 1/2 × 16 1/4 × 7"
Interior: 14 × 15 3/4 × 6 3/4"
Price
$684.53
Log in for availability
Case of 5
Minimum order quantity is 1
Product Details & Documents
These wire mesh bins offer a nickel-chrome finish and a 3/8” x 3/8” grid pattern allowing complete visibility of the bin’s contents.
  • Unique mesh pattern provides a dust and dirt free storage environment
  • Rear hanger allows them to hang from any louvered wall panel or rail system
Strong design allows superior hanging capacity for part storage where hanging is vital. A rear hanger allows the bin to hang from any louvered panel or rail system and optional side hanger which allows bins to stack one on top of the other.
Open hopper design allows for easy picking while allowing visual access to stored content. Two optional label holders are available for all Mesh Bins. Chrome color.
Specifications
  • Dimensions:
    Exterior: 14 1/2 × 16 1/4 × 7"
    Interior: 14 × 15 3/4 × 6 3/4"
Accessories
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Wire Divider, Chrome
    Wire Divider, Chrome
    10804-212
    QMB510C
    Log in for availability
    Each - $11.84
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Side Hanger, Chrome
    Side Hanger, Chrome
    10804-204
    QMB530C/QMB535C
    Log in for availability
    Pack of 1 - $9.09
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Side Hanger, Chrome
    Side Hanger, Chrome
    10804-206
    QMB539C
    Log in for availability
    Pack of 1 - $9.82
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Wire Divider, Chrome
    Wire Divider, Chrome
    10804-218
    QMB539C
    Log in for availability
    Each - $21.28
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Wire Divider, Chrome
    Wire Divider, Chrome
    10804-214
    QMB520C
    Log in for availability
    Each - $14.10
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Wire Divider, Chrome
    Wire Divider, Chrome
    10804-222
    QMB560C/QMB570C
    Log in for availability
    Each - $37.27
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Side Hanger, Chrome
    Side Hanger, Chrome
    10804-202
    QMB520C
    Log in for availability
    Pack of 1 - $8.75
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Wire Divider, Chrome
    Wire Divider, Chrome
    10804-216
    QMB530C/QMB535C
    Log in for availability
    Each - $20.21
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Wire Divider, Chrome
    Wire Divider, Chrome
    10804-220
    QMB540C/QMB550C
    Log in for availability
    Each - $24.48
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Side Hanger, Chrome
    Side Hanger, Chrome
    10804-208
    QMB540C/QMB550C
    Log in for availability
    Pack of 1 - $9.54
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Side Hanger, Chrome
    Side Hanger, Chrome
    10804-210
    QMB560C/QMB570C
    Log in for availability
    Each - $12.22