Order Entry
United States
Orders LinkContactUsLinkComponent
pH Indicator Papers

pH Indicator Papers

pH Indicator Papers, Litmus Red, Laboratory, Strips, Strips, Laboratory
Catalog # 470220-896
Supplier #:  190-12V-100
pH Indicator Papers
Catalog # 470220-896
pH Indicator Papers
pH Indicator Papers

pH Indicator Papers, Litmus Red, Laboratory, Strips, Strips, Laboratory

Price
$1.60
Log in for availability
Each Pack of 12
Product Details & Documents

pH Test Papers in your Choice of Two Indicator Colors.

  • Easy to read
  • Vial of 100

Litmus red paper changes to blue at pH 8.1. Color comparison chart not included.

Specifications
  • Description:
    pH Indicator Papers, Litmus Red, Laboratory, Strips
  • Grade:
    Laboratory
  • Form:
    Strips
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.