Orders LinkContactUsLinkComponent
Various Replacement Parts, Sciencix
Various Replacement Parts, Sciencix
Mixer Assembly, 100 μl, ACQUITY H-Class QSM, WATE, 700005119
Catalog # 103012-030
Marketsource
Supplier:   SCIENCIX
Supplier #:  CTS-A11376
Various Replacement Parts, Sciencix
Catalog # 103012-030
Supplier:  SCIENCIX
Marketsource
Various Replacement Parts, Sciencix
Various Replacement Parts, Sciencix
Mixer Assembly, 100 μl, ACQUITY H-Class QSM, WATE, 700005119
Price
$236.95
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
  • Ensures your system is working efficiently
  • Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
  • Description:
    Mixer Assembly, 100 μl
  • OEM manufacturer:
    WATE
  • OEM equivalent:
    700005119
  • Model:
    ACQUITY H-Class QSM