Various Replacement Parts, Sciencix
Primary Inlet CV Filter, Stainless Steel, ACQUITY I-Class BSM, ACQUITY Isocratic Solvent Mgr, ACQUITY M-Class µASM, ACQUITY M-Class µBSM, ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM, nanoACQUITY UPLC BSM/ASM, WATE, 289003547
Catalog # 103011-996
Marketsource
Supplier: SCIENCIX
: CTS-A11123
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Primary Inlet CV Filter, Stainless Steel
- OEM manufacturer:WATE
- OEM equivalent:289003547
- Model:ACQUITY I-Class BSM, ACQUITY Isocratic Solvent Mgr, ACQUITY M-Class µASM, ACQUITY M-Class µBSM, ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM, nanoACQUITY UPLC BSM/ASM