Orders LinkContactUsLinkComponent
Various Replacement Parts, Sciencix
Various Replacement Parts, Sciencix
Solvent Reservoir Filter, 10 µm, 600, 610, 510, 515, 1515, 1525, 1525EF, 1525 Micro, 2535, 2545, 2555, 2690, 2690D, 2695, 2695D, 2700, Acquity, Acquity UPLC BSM, Alliance, LC Module 1, WATE, WAT025531
Catalog # 103011-944
Marketsource
Supplier:   SCIENCIX
Supplier #:  CTS-5531
Various Replacement Parts, Sciencix
Catalog # 103011-944
Supplier:  SCIENCIX
Marketsource
Various Replacement Parts, Sciencix
Various Replacement Parts, Sciencix
Solvent Reservoir Filter, 10 µm, 600, 610, 510, 515, 1515, 1525, 1525EF, 1525 Micro, 2535, 2545, 2555, 2690, 2690D, 2695, 2695D, 2700, Acquity, Acquity UPLC BSM, Alliance, LC Module 1, WATE, WAT025531
Price
$58.17
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
  • Ensures your system is working efficiently
  • Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
  • Description:
    Solvent Reservoir Filter, 10 µm
  • OEM manufacturer:
    WATE
  • OEM equivalent:
    WAT025531
  • Model:
    600, 610, 510, 515, 1515, 1525, 1525EF, 1525 Micro, 2535, 2545, 2555, 2690, 2690D, 2695, 2695D, 2700, Acquity, Acquity UPLC BSM, Alliance, LC Module 1