Various Replacement Parts, Sciencix
Pre-Column Filter Assembly, 600, 712, 717, 2690, 2690D, 2695, 2695D, 2790, 2795, 2796, Alliance, LC Module 1, WATE, WAT035190
Catalog # 103011-866
Marketsource
Supplier: SCIENCIX
Supplier #: CTS-10527
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Pre-Column Filter Assembly
- OEM manufacturer:WATE
- OEM equivalent:WAT035190
- Model:600, 712, 717, 2690, 2690D, 2695, 2695D, 2790, 2795, 2796, Alliance, LC Module 1