Orders LinkContactUsLinkComponent
Various Replacement Parts, Sciencix
Various Replacement Parts, Sciencix
Line Filter Assembly, i-Series, LC-10ADvp, LC-10AT, LC-10ATvp, LC-20AD/AB, LC-20ADXR, LC-20AR, LC-20AT, LC-30ADSF, LC-2010, SHIM, 228-35871-96
Catalog # 103011-776
Marketsource
Supplier:   SCIENCIX
Supplier #:  CTS-A11421
Various Replacement Parts, Sciencix
Catalog # 103011-776
Supplier:  SCIENCIX
Marketsource
Various Replacement Parts, Sciencix
Various Replacement Parts, Sciencix
Line Filter Assembly, i-Series, LC-10ADvp, LC-10AT, LC-10ATvp, LC-20AD/AB, LC-20ADXR, LC-20AR, LC-20AT, LC-30ADSF, LC-2010, SHIM, 228-35871-96
Price
$87.89
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
  • Ensures your system is working efficiently
  • Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
  • Description:
    Line Filter Assembly
  • OEM manufacturer:
    SHIM
  • OEM equivalent:
    228-35871-96
  • Model:
    i-Series, LC-10ADvp, LC-10AT, LC-10ATvp, LC-20AD/AB, LC-20ADXR, LC-20AR, LC-20AT, LC-30ADSF, LC-2010