Various Replacement Parts, Sciencix
Capillary, 0.17ID×150L mm, 1100, 1200, 1260, AGLS, 5021-1817
Catalog # 103011-682
Marketsource
Supplier: SCIENCIX
Supplier #: CTS-11457
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Capillary, 0.17ID×150L mm
- OEM manufacturer:AGLS
- OEM equivalent:5021-1817
- Model:1100, 1200, 1260